Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rps28
Entrez Gene ID
100755309
Gene Name
ribosomal protein S28
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614895: 391812 - 393658 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651132.1 XP_007649322.1 3 - 4 220 67

View Full Product Details