Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dkkl1
Entrez Gene ID
100771803
Gene Name
dickkopf-like 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614570: 550152 - 554459 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508644.2 XP_003508692.1 3 - 4 388 79

View Full Product Details