Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC100774508
Entrez Gene ID
100774508
Gene Name
chromosome unknown open reading frame, human C11orf35
Gene Aliases
I79_017962
Chromosome Location
Chr.Un|NW_003614763: 65132 - 71058 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509753.2 XP_003509801.1 12 - 13 1573 72

View Full Product Details