Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asxl1
Entrez Gene ID
100761070
Gene Name
additional sex combs like 1 (Drosophila)
Gene Aliases
I79_008871
Chromosome Location
Chr.Un|NW_003613890: 641120 - 708035 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502341.2 XP_003502389.1 10 - 11 1295 81

View Full Product Details