Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scarf1
Entrez Gene ID
100767435
Gene Name
scavenger receptor class F, member 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616958: 25865 - 39606 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514778.2 XP_003514826.1 7 - 8 1584 74

View Full Product Details