Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zic1
Entrez Gene ID
100764121
Gene Name
Zic family member 1
Gene Aliases
I79_000875
Chromosome Location
Chr.Un|NW_003613597: 350720 - 353800 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495645.1 XP_003495693.1 2 - 3 631 57

View Full Product Details