Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fibin
Entrez Gene ID
100760017
Gene Name
fin bud initiation factor homolog (zebrafish)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614250: 142882 - 144738 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506508.2 XP_003506556.1 1 - 1 720 75

View Full Product Details