Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syngr3
Entrez Gene ID
100767759
Gene Name
synaptogyrin 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613852: 1605856 - 1610593 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501545.2 XP_003501593.2 3 - 4 468 59

View Full Product Details