Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Has3
Entrez Gene ID
100757895
Gene Name
hyaluronan synthase 3
Gene Aliases
I79_011328
Chromosome Location
Chr.Un|NW_003614050: 133851 - 151000 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504303.2 XP_003504351.1 2 - 3 719 59
XM_007644837.1 XP_007643027.1 1 - 2 3135 59

View Full Product Details