Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Siglec14
Entrez Gene ID
103164379
Gene Name
sialic acid binding Ig-like lectin 14
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615616: 217170 - 220982 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653253.1 XP_007651443.1 4 - 5 304 73

View Full Product Details