Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rtdr1
Entrez Gene ID
100767963
Gene Name
rhabdoid tumor deletion region gene 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614107: 861188 - 935038 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645454.1 XP_007643644.1 5 - 6 783 67

View Full Product Details