Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zpr1
Entrez Gene ID
100773173
Gene Name
ZPR1 zinc finger
Gene Aliases
I79_019552, Zfp259
Chromosome Location
Chr.Un|NW_003615029: 321580 - 330138 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511092.2 XP_003511140.1 10 - 11 1027 73
XM_007651653.1 XP_007649843.1 10 - 11 1027 73
GenBank mRNA JI877779.1 - 1 - 2 88 73
JP040500.1 - 1 - 2 88 73

View Full Product Details