Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tlr10
Entrez Gene ID
100759685
Gene Name
toll-like receptor 10
Gene Aliases
I79_025190
Chromosome Location
Chr.Un|NW_003618519: 28901 - 31330 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515569.1 XP_003515617.1 1 - 1 2020 108

View Full Product Details