Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zrsr2
Entrez Gene ID
100769034
Gene Name
zinc finger (CCCH type), RNA-binding motif and serine/arginine rich 2
Gene Aliases
I79_020059
Chromosome Location
Chr.Un|NW_003615140: 194385 - 221454 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652072.1 XP_007650262.1 10 - 11 1269 62

View Full Product Details