Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pld6
Entrez Gene ID
100766800
Gene Name
phospholipase D family, member 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613960: 1050002 - 1051147 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643973.1 XP_007642163.1 1 - 2 92 61

View Full Product Details