Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Igsf22
Entrez Gene ID
103164228
Gene Name
immunoglobulin superfamily, member 22
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614690: 387181 - 412473 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649932.1 XP_007648122.1 3 - 4 331 69

View Full Product Details