Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prdm6
Entrez Gene ID
100773424
Gene Name
PR domain containing 6
Gene Aliases
I79_011131
Chromosome Location
Chr.Un|NW_003614035: 383146 - 522500 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504132.2 XP_003504180.2 7 - 8 1259 60

View Full Product Details