Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc30a10
Entrez Gene ID
100774627
Gene Name
solute carrier family 30, member 10
Gene Aliases
I79_000087
Chromosome Location
Chr.Un|NW_003613581: 3443458 - 3473139 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003494972.1 XP_003495020.1 3 - 4 690 94

View Full Product Details