Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Odf3l2
Entrez Gene ID
100761563
Gene Name
outer dense fiber of sperm tails 3-like 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613904: 421503 - 427134 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502552.1 XP_003502600.1 2 - 3 235 60

View Full Product Details