Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc11
Entrez Gene ID
100763503
Gene Name
coiled-coil domain containing 11
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614193: 25125 - 70722 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007646313.1 XP_007644503.1 3 - 4 484 81

View Full Product Details