Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serf2
Entrez Gene ID
100754083
Gene Name
small EDRK-rich factor 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613677: 853915 - 855507 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007621473.1 XP_007619663.1 2 - 3 203 101

View Full Product Details