Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rtn4rl1
Entrez Gene ID
100772121
Gene Name
reticulon 4 receptor-like 1
Gene Aliases
I79_023705
Chromosome Location
Chr.Un|NW_003616642: 43692 - 47147 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654593.1 XP_007652783.1 1 - 2 50 73

View Full Product Details