Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atox1
Entrez Gene ID
100760386
Gene Name
antioxidant 1 copper chaperone
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613733: 1448084 - 1461230 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499376.2 XP_003499424.1 3 - 4 354 95
GenBank mRNA JI879305.1 - 3 - 4 349 95
JP041124.1 - 3 - 4 349 95

View Full Product Details