Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ggt6
Entrez Gene ID
100758668
Gene Name
gamma-glutamyltransferase 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614234: 521978 - 524689 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506378.2 XP_003506426.1 3 - 4 457 61

View Full Product Details