Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb46
Entrez Gene ID
100762624
Gene Name
zinc finger and BTB domain containing 46
Gene Aliases
I79_009896
Chromosome Location
Chr.Un|NW_003613943: 1354788 - 1394433 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503138.2 XP_003503186.1 3 - 4 1397 79

View Full Product Details