Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rpusd2
Entrez Gene ID
100752640
Gene Name
RNA pseudouridylate synthase domain containing 2
Gene Aliases
I79_016219
Chromosome Location
Chr.Un|NW_003614523: 204930 - 209496 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508317.2 XP_003508365.1 1 - 2 664 73

View Full Product Details