Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc22
Entrez Gene ID
100770201
Gene Name
tetratricopeptide repeat domain 22
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613599: 3290813 - 3308729 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495740.1 XP_003495788.1 2 - 3 621 90

View Full Product Details