Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp36l1
Entrez Gene ID
100759015
Gene Name
ZFP36 ring finger protein-like 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613686: 1385643 - 1390896 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498261.2 XP_003498309.1 1 - 2 211 108

View Full Product Details