Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pfdn2
Entrez Gene ID
100766028
Gene Name
prefoldin subunit 2
Gene Aliases
I79_014378
Chromosome Location
Chr.Un|NW_003614301: 172327 - 180634 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506857.2 XP_003506905.1 2 - 3 206 72
GenBank mRNA JI893896.1 - 2 - 3 201 72

View Full Product Details