Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map1s
Entrez Gene ID
100772627
Gene Name
microtubule-associated protein 1S
Gene Aliases
I79_005123, Mtap1s
Chromosome Location
Chr.Un|NW_003613716: 1182168 - 1191264 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007638529.1 XP_007636719.1 3 - 4 411 95

View Full Product Details