Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Frmpd3
Entrez Gene ID
100760412
Gene Name
FERM and PDZ domain containing 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614343: 353164 - 396917 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647825.1 XP_007646015.1 2 - 3 162 108

View Full Product Details