Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rps23
Entrez Gene ID
100772739
Gene Name
ribosomal protein S23
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613994: 849525 - 851303 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503725.2 XP_003503773.1 2 - 3 238 68
GenBank mRNA JI875884.1 - 1 - 2 407 68
JP044665.1 - 2 - 3 221 68

View Full Product Details