Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scml1
Entrez Gene ID
103160845
Gene Name
sex comb on midleg-like 1 (Drosophila)
Gene Aliases
I79_011232
Chromosome Location
Chr.Un|NW_003614044: 655344 - 667196 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644743.1 XP_007642933.1 4 - 5 308 103

View Full Product Details