Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dfnb31
Entrez Gene ID
100754986
Gene Name
deafness, autosomal recessive 31
Gene Aliases
Whrn
Chromosome Location
Chr.Un|NW_003613973: 156401 - 252262 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503466.2 XP_003503514.1 9 - 10 1194 67

View Full Product Details