Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asb4
Entrez Gene ID
100763121
Gene Name
ankyrin repeat and SOCS box containing 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614269: 724416 - 760822 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506650.1 XP_003506698.1 3 - 4 975 73

View Full Product Details