Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc15a3
Entrez Gene ID
100775005
Gene Name
solute carrier family 15 (oligopeptide transporter), member 3
Gene Aliases
I79_021948
Chromosome Location
Chr.Un|NW_003615705: 124723 - 141323 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513019.2 XP_003513067.1 1 - 2 1278 67

View Full Product Details