Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnj6
Entrez Gene ID
100763599
Gene Name
potassium inwardly-rectifying channel, subfamily J, member 6
Gene Aliases
I79_014889
Chromosome Location
Chr.Un|NW_003614346: 523601 - 588152 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647832.1 XP_007646022.1 1 - 2 899 91

View Full Product Details