Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnajc12
Entrez Gene ID
100758554
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 12
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613826: 556913 - 572871 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641729.1 XP_007639919.1 4 - 5 626 54

View Full Product Details