Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kcns3
Entrez Gene ID
100755672
Gene Name
potassium voltage-gated channel, delayed-rectifier, subfamily S, member 3
Gene Aliases
I79_009771
Chromosome Location
Chr.Un|NW_003613936: 838915 - 840390 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643673.1 XP_007641863.1 1 - 1 1425 73

View Full Product Details