Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vhl
Entrez Gene ID
100751916
Gene Name
von Hippel-Lindau tumor suppressor, E3 ubiquitin protein ligase
Gene Aliases
I79_002351
Chromosome Location
Chr.Un|NW_003613637: 681294 - 690240 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654374.1 XP_007652564.1 1 - 2 280 262

View Full Product Details