Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppm1e
Entrez Gene ID
103163832
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent, 1E
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614004: 624629 - 654173 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644384.1 XP_007642574.1 2 - 3 313 62

View Full Product Details