Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spty2d1
Entrez Gene ID
100752246
Gene Name
SPT2, Suppressor of Ty, domain containing 1 (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614690: 317959 - 337784 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509345.2 XP_003509393.1 1 - 2 251 64
GenBank mRNA JI865484.1 - 1 - 2 155 64
JP040947.1 - 1 - 2 155 64

View Full Product Details