Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam3b
Entrez Gene ID
100764355
Gene Name
family with sequence similarity 3, member B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613905: 527010 - 553299 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502640.2 XP_003502688.1 7 - 8 624 70

View Full Product Details