Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hoxd4
Entrez Gene ID
103161712
Gene Name
homeobox D4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613688: 1409341 - 1417270 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007626360.1 XP_007624550.1 6 - 7 900 59

View Full Product Details