Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd7
Entrez Gene ID
100755680
Gene Name
ankyrin repeat domain 7
Gene Aliases
I79_012450
Chromosome Location
Chr.Un|NW_003614134: 32315 - 46358 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505281.1 XP_003505329.1 5 - 6 772 85

View Full Product Details