Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rem2
Entrez Gene ID
100764771
Gene Name
RAS (RAD and GEM)-like GTP binding 2
Gene Aliases
I79_013397
Chromosome Location
Chr.Un|NW_003614213: 565700 - 569111 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007646601.1 XP_007644791.1 3 - 4 505 79

View Full Product Details