Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zc3h12b
Entrez Gene ID
100774530
Gene Name
zinc finger CCCH-type containing 12B
Gene Aliases
I79_023021
Chromosome Location
Chr.Un|NW_003616217: 54710 - 81898 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654158.1 XP_007652348.1 1 - 2 755 118

View Full Product Details