Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gabrr1
Entrez Gene ID
100769153
Gene Name
gamma-aminobutyric acid (GABA) A receptor, rho 1
Gene Aliases
I79_000683
Chromosome Location
Chr.Un|NW_003613592: 897401 - 913477 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641080.1 XP_007639270.1 5 - 6 514 98

View Full Product Details