Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nefm
Entrez Gene ID
100773854
Gene Name
neurofilament, medium polypeptide
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613613: 3862819 - 3867328 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496187.2 XP_003496235.1 1 - 2 1071 70

View Full Product Details