Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il17c
Entrez Gene ID
100767127
Gene Name
interleukin 17C
Gene Aliases
I79_000234
Chromosome Location
Chr.Un|NW_003613583: 1114961 - 1120213 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653032.1 XP_007651222.1 2 - 3 1140 58

View Full Product Details